KEY458251833 Key Organics / BIONET

(3E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one

  • MDL number MFCD00051608
  • Cas Number : 366815-43-2
  • Molecular Weight : 198.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81929

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/6H-022_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart