KEY458251854 Key Organics / BIONET

3-methyl-3,4-dihydro-1,2,3-benzotriazin-4-one

  • MDL number MFCD04125779
  • Cas Number : 22305-44-8
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82483

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6K-546S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart