KEY458251885 Key Organics / BIONET

N'-(5-chloropentanoyl)-4-fluorobenzohydrazide

  • MDL number MFCD28976603
  • Cas Number : 1823188-46-0
  • Molecular Weight : 272.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/529173

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AD-0101_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart