KEY458251910 Key Organics / BIONET

methyl (2E)-3-(2-fluorophenyl)prop-2-enoate

  • MDL number MFCD03102529
  • Cas Number : 104201-65-2
  • Molecular Weight : 180.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83949

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6T-0039_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart