KEY458251923 Key Organics / BIONET

7,8-dimethoxy-2,3-dihydro-1H-3-benzazepin-2-one

  • MDL number MFCD02091565
  • Cas Number : 73942-87-7
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84426

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6X-0944_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart