KEY458251937 Key Organics / BIONET

3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD00231871
  • Cas Number : 318951-60-9
  • Molecular Weight : 161.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile
Inchi Code 1S/C5H2F3N3/c6-5(7,8)4-3(1-9)2-10-11-4/h2H,(H,10,11)
Inchi Key HNIRANRLHUHUOS-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85358

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/7H-076_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart