KEY458251941 Key Organics / BIONET

1-(4-chlorophenyl)-2-methanesulfonylethan-1-one

  • MDL number MFCD00202209
  • Cas Number : 24437-48-7
  • Molecular Weight : 232.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85566

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7H-573S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart