KEY458252001 Key Organics / BIONET

3-bromo-8-methylimidazo[1,2-a]pyridine

  • MDL number MFCD06496194
  • Cas Number : 866135-66-2
  • Molecular Weight : 211.062

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88147

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7X-0845_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart