KEY458252003 Key Organics / BIONET

(2E)-3-(cyclohexylcarbamoyl)prop-2-enoic acid

  • MDL number MFCD00086001
  • Cas Number : 113467-97-3
  • Molecular Weight : 197.234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88151

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7X-0852_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart