KEY458252018 Key Organics / BIONET

5-(pyridin-2-yl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD00139078
  • Cas Number : 3690-47-9
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(2-pyridinyl)-1,3,4-oxadiazole-2(3H)-thione
Inchi Code 1S/C7H5N3OS/c12-7-10-9-6(11-7)5-3-1-2-4-8-5/h1-4H,(H,10,12)
Inchi Key KXNWRPICZLNODY-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88476

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/8D-050_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart