KEY458252031 Key Organics / BIONET

1-[(4-bromophenyl)sulfanyl]-4-nitrobenzene

  • MDL number MFCD00172327
  • Cas Number : 21969-12-0
  • Molecular Weight : 310.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[(4-bromophenyl)sulfanyl]-4-nitrobenzene
Inchi Code 1S/C12H8BrNO2S/c13-9-1-5-11(6-2-9)17-12-7-3-10(4-8-12)14(15)16/h1-8H
Inchi Key XWPBPDJTKDECOY-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88930

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/8G-348S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart