KEY458252073 Key Organics / BIONET

4-(chloromethyl)-7-hydroxy-8-methyl-2H-chromen-2-one

  • MDL number MFCD03937668
  • Cas Number : 163684-50-2
  • Molecular Weight : 224.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91049

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8W-0243_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart