KEY458252074 Key Organics / BIONET

2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylic acid

  • MDL number MFCD04126131
  • Cas Number : 149771-23-3
  • Molecular Weight : 207.112

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91058

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8W-0262_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart