KEY458252079 Key Organics / BIONET

6-hydrazinyl-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD00091860
  • Molecular Weight : 170.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91142

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8W-0876_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart