KEY458252080 Key Organics / BIONET

1-(2,2,2-trifluoroacetyl)piperidine-4-carboxylic acid

  • MDL number MFCD03840837
  • Cas Number : 126501-70-0
  • Molecular Weight : 225.167

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91148

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8W-0884_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart