KEY458252087 Key Organics / BIONET

2-cyano-N-phenylacetamide

  • MDL number MFCD00019840
  • Cas Number : 621-03-4
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 198-202
Iupac Name 2-cyano-N-phenylacetamide
Inchi Code 1S/C9H8N2O/c10-7-6-9(12)11-8-4-2-1-3-5-8/h1-5H,6H2,(H,11,12)
Inchi Key XCTQPMCULSZKLT-UHFFFAOYSA-N

Description

Melting Point 198-202
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91413

Safety Information

signalword Warning
pcodes P260,P270,P280,P301 + P312 + P330,P302 + P352,P304 + P340,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/9B-046_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart