KEY458252116 Key Organics / BIONET

3-acetamidothiophene-2-carboxylic acid

  • MDL number MFCD00101396
  • Cas Number : 50901-18-3
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(acetylamino)-2-thiophenecarboxylic acid
Inchi Code 1S/C7H7NO3S/c1-4(9)8-5-2-3-12-6(5)7(10)11/h2-3H,1H3,(H,8,9)(H,10,11)
Inchi Key LLKLTQJOEPWBOE-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92943

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9M-523S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart