KEY458252117 Key Organics / BIONET

pyrimidine-4-carbohydrazide

  • MDL number MFCD06738862
  • Cas Number : 39513-54-7
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-pyrimidinecarbohydrazide
Inchi Code 1S/C5H6N4O/c6-9-5(10)4-1-2-7-3-8-4/h1-3H,6H2,(H,9,10)
Inchi Key NWTQBXDLKZTWAH-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93088

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9N-072_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart