KEY458252129 Key Organics / BIONET

3-{[(tert-butoxy)carbonyl]amino}-3-(naphthalen-1-yl)propanoic acid

  • MDL number MFCD02090687
  • Cas Number : 268209-95-6
  • Molecular Weight : 315.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(tert-butoxycarbonyl)-3-(1-naphthyl)-beta-alanine
Inchi Code 1S/C18H21NO4/c1-18(2,3)23-17(22)19-15(11-16(20)21)14-10-6-8-12-7-4-5-9-13(12)14/h4-10,15H,11H2,1-3H3,(H,19,22)(H,20,21)
Inchi Key YDSAGJMVVUBPOK-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93730

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9R-0662_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart