KEY458252130 Key Organics / BIONET

4-(4-methoxyphenyl)-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine

  • MDL number MFCD01839591
  • Cas Number : 4875-49-4
  • Molecular Weight : 229.283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93775

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-1075_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart