KEY458252148 Key Organics / BIONET

5-(3-bromophenyl)-1,3-oxazole

  • MDL number MFCD05668970
  • Cas Number : 243455-57-4
  • Molecular Weight : 224.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(3-bromophenyl)-1,3-oxazole
Inchi Code 1S/C9H6BrNO/c10-8-3-1-2-7(4-8)9-5-11-6-12-9/h1-6H
Inchi Key GUYZQMXSOVOSIC-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94248

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9X-0851_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart