KEY458252149 Key Organics / BIONET

5-(3-nitrophenyl)-1,3-oxazole

  • MDL number MFCD00085147
  • Cas Number : 89808-77-5
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(3-nitrophenyl)-1,3-oxazole
Inchi Code 1S/C9H6N2O3/c12-11(13)8-3-1-2-7(4-8)9-5-10-6-14-9/h1-6H
Inchi Key ALXACHUFQSLYBV-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94255

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9X-0862_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart