KEY458252164 Key Organics / BIONET

methyl 4'-methyl-[1,1'-biphenyl]-4-carboxylate

  • MDL number MFCD05979484
  • Cas Number : 49742-56-5
  • Molecular Weight : 226.275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/529179

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2071_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart