KEY458252167 Key Organics / BIONET

[4-(9H-carbazol-9-yl)phenyl]boronic acid

  • MDL number MFCD13176534
  • Cas Number : 419536-33-7
  • Molecular Weight : 287.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(9H-carbazol-9-yl)phenylboronic acid
Inchi Code 1S/C18H14BNO2/c21-19(22)13-9-11-14(12-10-13)20-17-7-3-1-5-15(17)16-6-2-4-8-18(16)20/h1-12,21-22H
Inchi Key JGAVTCVHDMOQTJ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store under N2
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/529182

Safety Information

signalword Warning
pcodes P422, P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3170_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart