KEY458252174 Key Organics / BIONET

3-(4-bromophenyl)propan-1-ol

  • MDL number MFCD09028724
  • Cas Number : 25574-11-2
  • Molecular Weight : 215.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-bromophenyl)-1-propanol
Inchi Code 1S/C9H11BrO/c10-9-5-3-8(4-6-9)2-1-7-11/h3-6,11H,1-2,7H2
Inchi Key WODKXGCVVOOEIJ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/529189

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3177_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart