KEY458252178 Key Organics / BIONET

2-(thiophen-2-yl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD00022489
  • Cas Number : 32451-19-7
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-thienyl)-1,3-thiazolidine-4-carboxylic acid
Inchi Code 1S/C8H9NO2S2/c10-8(11)5-4-13-7(9-5)6-2-1-3-12-6/h1-3,5,7,9H,4H2,(H,10,11)
Inchi Key ZUDFHYZUQCNUJF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83376

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6P-124_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart