KEY458252182 Key Organics / BIONET

4',5-dimethyl-[1,1'-biphenyl]-2-carboxylic acid

  • MDL number MFCD23684901
  • Cas Number : 474519-95-4
  • Molecular Weight : 226.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/529197

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3186_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart