KEY458252186 Key Organics / BIONET

2-(2-iodo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD17215809
  • Cas Number : 1256781-69-7
  • Molecular Weight : 360

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/529201

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3190_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart