KEY458252192 Key Organics / BIONET

4-methyl-2-(1H-pyrazol-3-yl)phenol

  • MDL number MFCD03424535
  • Cas Number : 57148-86-4
  • Molecular Weight : 174.203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94899

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BA-0843_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart