KEY458252205 Key Organics / BIONET

2-methyl-1H-indol-4-ol

  • MDL number MFCD00152133
  • Cas Number : 35320-67-3
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-1H-indol-4-ol
Inchi Code 1S/C9H9NO/c1-6-5-7-8(10-6)3-2-4-9(7)11/h2-5,10-11H,1H3
Inchi Key XBVSGEGNQZAQPM-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95870

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CG-0507_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart