KEY458252249 Key Organics / BIONET

1-[(4-bromo-2-fluorophenyl)methyl]pyrrolidine

  • MDL number MFCD09749655
  • Cas Number : 283173-83-1
  • Molecular Weight : 258.134

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/358466

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FG-0402_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart