KEY458252266 Key Organics / BIONET

6-oxo-1,6-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD00052805
  • Cas Number : 37972-69-3
  • Molecular Weight : 140.098

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97081

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FG-0444_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart