KEY458252337 Key Organics / BIONET

6-bromo-4-oxo-4H-chromene-3-carbonitrile

  • MDL number MFCD00191842
  • Cas Number : 52817-13-7
  • Molecular Weight : 250.051

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222 - 224
Purity 95
Storage temp. Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/403201

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FS-1292_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart