KEY458252463 Key Organics / BIONET

2-(1H-1,3-benzodiazol-2-yl)acetic acid

  • MDL number MFCD01102656
  • Cas Number : 13570-08-6
  • Molecular Weight : 176.175

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/403770

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P312,P402 + P404,P411
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FS-1876_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart