KEY458252475 Key Organics / BIONET

{4-[(morpholin-4-yl)methyl]phenyl}methanamine

  • MDL number MFCD05664410
  • Cas Number : 91271-84-0
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(4-morpholinylmethyl)phenyl]methanamine
Inchi Code 1S/C12H18N2O/c13-9-11-1-3-12(4-2-11)10-14-5-7-15-8-6-14/h1-4H,5-10,13H2
Inchi Key OTMQHFNPBREFIL-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/403942

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-2055_SDS.pdf
hcodes H302,H312,H314,H315,H319,H332,H335
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart