KEY458252520 Key Organics / BIONET

benzyl 3-hydroxypiperidine-1-carboxylate

  • MDL number MFCD03839903
  • Cas Number : 95798-22-4
  • Molecular Weight : 235.283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/404178

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FS-2293_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart