KEY458252561 Key Organics / BIONET

[(3S,4R)-4-(4-fluorophenyl)-1-methylpiperidin-3-yl]methanol

  • MDL number MFCD09842552
  • Cas Number : 105812-81-5
  • Molecular Weight : 223.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [(3S,4R)-4-(4-fluorophenyl)-1-methyl-3-piperidinyl]methanol
Inchi Code 1S/C13H18FNO/c1-15-7-6-13(11(8-15)9-16)10-2-4-12(14)5-3-10/h2-5,11,13,16H,6-9H2,1H3/t11-,13-/m0/s1
Inchi Key CXRHUYYZISIIMT-AAEUAGOBSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/404443

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-2563_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart