KEY458252631 Key Organics / BIONET

ethyl 5-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD01006731
  • Cas Number : 3284-51-3
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-methyl-1H-pyrrole-2-carboxylate
Inchi Code 1S/C8H11NO2/c1-3-11-8(10)7-5-4-6(2)9-7/h4-5,9H,3H2,1-2H3
Inchi Key RIUNZXNCMZMRMP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/404918

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-3047_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart