KEY458252644 Key Organics / BIONET

5-chloro-2-hydroxybenzamide

  • MDL number MFCD00007980
  • Cas Number : 7120-43-6
  • Molecular Weight : 171.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/405017

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FS-3148_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart