KEY458252663 Key Organics / BIONET

3-benzoylpyridine

  • MDL number MFCD00006394
  • Cas Number : 5424-19-1
  • Molecular Weight : 183.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/405133

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FS-3267_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart