KEY458252687 Key Organics / BIONET

3-(piperidin-1-yl)phenol

  • MDL number MFCD00053001
  • Cas Number : 27292-50-8
  • Molecular Weight : 177.247

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/405357

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FS-3499_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart