KEY458252703 Key Organics / BIONET

4-(piperazin-1-yl)phenol

  • MDL number MFCD00066156
  • Cas Number : 56621-48-8
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 220-225
Iupac Name 4-(1-piperazinyl)phenol
Inchi Code 1S/C10H14N2O/c13-10-3-1-9(2-4-10)12-7-5-11-6-8-12/h1-4,11,13H,5-8H2
Inchi Key GPEOAEVZTOQXLG-UHFFFAOYSA-N

Description

Melting Point 220-225
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/405627

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301 + P312 + P330,P302 + P352,P302 + P352 + P312,P304 + P340 + P311,P305 + P351 + P338,P312,P332 + P313,P337 + P313,P362,P402 + P404,P403 + P233,P405,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-3772_SDS.pdf
hcodes H302,H315,H319,H332,H335
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart