KEY458252716 Key Organics / BIONET

tert-butyl 6-chloropyridine-3-carboxylate

  • MDL number MFCD08277247
  • Cas Number : 115309-57-4
  • Molecular Weight : 213.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 6-chloronicotinate
Inchi Code 1S/C10H12ClNO2/c1-10(2,3)14-9(13)7-4-5-8(11)12-6-7/h4-6H,1-3H3
Inchi Key OFRLMTIBUAXBGV-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97817

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/HE-0017_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart