KEY458252718 Key Organics / BIONET

5-chlorofuran-2-carboxylic acid

  • MDL number MFCD00093272
  • Cas Number : 618-30-4
  • Molecular Weight : 146.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-furoic acid
Inchi Code 1S/C5H3ClO3/c6-4-2-1-3(9-4)5(7)8/h1-2H,(H,7,8)
Inchi Key NNTBDUTXMIMRKK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store under N2 at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97955

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/HG-0015_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart