KEY458252719 Key Organics / BIONET

methyl 4-chloro-3-iodobenzoate

  • MDL number MFCD09263357
  • Cas Number : 365526-34-7
  • Molecular Weight : 296.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-chloro-3-iodobenzoate
Inchi Code 1S/C8H6ClIO2/c1-12-8(11)5-2-3-6(9)7(10)4-5/h2-4H,1H3
Inchi Key LWENYAHUBUWONE-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100870

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/KA-5035_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart