KEY458253602 Key Organics / BIONET

3,5-dimethylpyrazin-2-amine

  • MDL number MFCD00107172
  • Cas Number : 91678-81-8
  • Molecular Weight : 123.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-dimethyl-2-pyrazinamine
Inchi Code 1S/C6H9N3/c1-4-3-8-6(7)5(2)9-4/h3H,1-2H3,(H2,7,8)
Inchi Key GZZRWBRYYZGTNW-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/406779

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/PS-4172_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart