KEY458253767 Key Organics / BIONET

ethyl 3-(methylsulfanyl)-4-oxo-4,5,6,7-tetrahydro-2-benzothiophene-1-carboxylate

  • MDL number MFCD00085083
  • Cas Number : 168279-54-7
  • Molecular Weight : 270.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-(methylsulfanyl)-4-oxo-4,5,6,7-tetrahydro-2-benzothiophene-1-carboxylate
Inchi Code 1S/C12H14O3S2/c1-3-15-11(14)10-7-5-4-6-8(13)9(7)12(16-2)17-10/h3-6H2,1-2H3
Inchi Key BHVLZDJJLSBOHJ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/406989

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/PS-4388_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart