KEY458254044 Key Organics / BIONET

1-methyl-1H-indazol-4-ol

  • MDL number MFCD11869776
  • Cas Number : 144528-23-4
  • Molecular Weight : 148.165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/407376

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P422,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/PS-4791_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart