KEY458254244 Key Organics / BIONET

5-methoxypyridin-2-amine

  • MDL number MFCD07374873
  • Cas Number : 10167-97-2
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methoxy-2-pyridinamine
Inchi Code 1S/C6H8N2O/c1-9-5-2-3-6(7)8-4-5/h2-4H,1H3,(H2,7,8)
Inchi Key XJKJHILCYUUVSJ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store under N2 at -18
Physical Form Low Melting Solid,Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/407621

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/PS-5045_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart