KEY458256080 Key Organics / BIONET

prop-2-en-1-yl 3,5-diamino-1H-pyrazole-4-carboxylate

  • MDL number MFCD27996731
  • Cas Number : 1613191-73-3
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name allyl 3,5-diamino-1H-pyrazole-4-carboxylate
Inchi Code 1S/C7H10N4O2/c1-2-3-13-7(12)4-5(8)10-11-6(4)9/h2H,1,3H2,(H5,8,9,10,11)
Inchi Key QPTWDVFFLRWIAM-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/529551

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4915_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart